Assay Design



Gene Transcripts

Gene Symbol
Guca2a
Entrez Gene ID
100755664
Gene Name
guanylate cyclase activator 2A (guanylin)
Gene Aliases
Guanylin
Chromosome Location
Chr.Un|NW_003613862: 1620587 - 1622676 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501825.2 XP_003501873.2 1 - 2 343 85

View Full Product Details