Assay Design



Gene Transcripts

Gene Symbol
Nipal4
Entrez Gene ID
100761365
Gene Name
NIPA-like domain containing 4
Gene Aliases
I79_009447
Chromosome Location
Chr.Un|NW_003613914: 7436 - 26687 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502788.1 XP_003502836.1 4 - 5 530 65

View Full Product Details