Assay Design



Gene Transcripts

Gene Symbol
Tm2d2
Entrez Gene ID
100774168
Gene Name
TM2 domain containing 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613685: 2534888 - 2539563 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498230.2 XP_003498278.1 1 - 2 334 87
GenBank mRNA JI884409.1 - 1 - 2 164 87
JP058848.1 - 1 - 2 164 87

View Full Product Details