Assay Design



Gene Transcripts

Gene Symbol
Ifngr2
Entrez Gene ID
100768877
Gene Name
interferon gamma receptor 2 (interferon gamma transducer 1)
Gene Aliases
I79_000826
Chromosome Location
Chr.Un|NW_003613595: 4166740 - 4179991 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642060.1 XP_007640250.1 6 - 7 1157 84
XM_007642065.1 XP_007640255.1 6 - 7 1185 84
GenBank mRNA JI865507.1 - 5 - 6 775 84

View Full Product Details