Assay Design



Gene Transcripts

Gene Symbol
Magel2
Entrez Gene ID
100769128
Gene Name
MAGE-like 2
Gene Aliases
I79_016372
Chromosome Location
Chr.Un|NW_003614542: 146428 - 150331 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508428.1 XP_003508476.1 1 - 1 552 168

View Full Product Details