Assay Design



Gene Transcripts

Gene Symbol
Klk5
Entrez Gene ID
100756768
Gene Name
kallikrein-related peptidase 5
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614844: 73923 - 81784 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007650818.1 XP_007649008.1 1 - 2 72 76

View Full Product Details