Assay Design



Gene Transcripts

Gene Symbol
Iglon5
Entrez Gene ID
100754556
Gene Name
IgLON family member 5
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615616: 162106 - 167708 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653252.1 XP_007651442.1 6 - 7 706 79

View Full Product Details