Assay Design



Gene Transcripts

Gene Symbol
Fgf19
Entrez Gene ID
100758307
Gene Name
fibroblast growth factor 19
Gene Aliases
I79_018316
Chromosome Location
Chr.Un|NW_003614822: 272427 - 276155 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510066.1 XP_003510114.1 2 - 3 258 83

View Full Product Details