Assay Design



Gene Transcripts

Gene Symbol
Trmt5
Entrez Gene ID
100752861
Gene Name
tRNA methyltransferase 5
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615679: 165474 - 171938 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512971.2 XP_003513019.2 4 - 5 1546 82
XM_007653345.1 XP_007651535.1 4 - 5 1528 82
GenBank mRNA JI883895.1 - 1 - 2 658 82
JP063809.1 - 1 - 2 658 82

View Full Product Details