Assay Design



Gene Transcripts

Gene Symbol
Clic3
Entrez Gene ID
100763283
Gene Name
chloride intracellular channel 3
Gene Aliases
I79_025059
Chromosome Location
Chr.Un|NW_003618227: 34485 - 36315 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515491.2 XP_003515539.1 5 - 6 551 94

View Full Product Details