Assay Design



Gene Transcripts

Gene Symbol
Tubal3
Entrez Gene ID
100755907
Gene Name
tubulin, alpha-like 3
Gene Aliases
I79_000022
Chromosome Location
Chr.Un|NW_003613580: 4022397 - 4031656 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007608071.1 XP_007606261.1 3 - 4 457 84

View Full Product Details