Assay Design



Gene Transcripts

Gene Symbol
Eif2d
Entrez Gene ID
100759038
Gene Name
eukaryotic translation initiation factor 2D
Gene Aliases
I79_007610, Ligatin
Chromosome Location
Chr.Un|NW_003613838: 1735929 - 1752879 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501218.2 XP_003501266.1 14 - 15 1836 106
XM_007641906.1 XP_007640096.1 14 - 15 1672 106
XM_007641907.1 XP_007640097.1 13 - 14 1686 106
GenBank mRNA JP061130.1 - 9 - 10 1107 106

View Full Product Details