Assay Design



Gene Transcripts

Gene Symbol
Slc16a1
Entrez Gene ID
100775004
Gene Name
solute carrier family 16 (monocarboxylate transporter), member 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615675: 197979 - 208549 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512965.2 XP_003513013.1 1 - 2 275 104
GenBank mRNA JI887939.1 - 1 - 2 267 104
JP043039.1 - 1 - 2 267 104

View Full Product Details