Assay Design



Gene Transcripts

Gene Symbol
Slc16a5
Entrez Gene ID
100757367
Gene Name
solute carrier family 16 (monocarboxylate transporter), member 5
Gene Aliases
I79_000937
Chromosome Location
Chr.Un|NW_003613598: 413684 - 424094 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643055.1 XP_007641245.1 2 - 3 344 67

View Full Product Details