Assay Design



Gene Transcripts

Gene Symbol
Dok7
Entrez Gene ID
100765890
Gene Name
docking protein 7
Gene Aliases
I79_002032
Chromosome Location
Chr.Un|NW_003613629: 334856 - 357622 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496603.1 XP_003496651.1 1 - 2 217 61

View Full Product Details