Assay Design



Gene Transcripts

Gene Symbol
Psmg2
Entrez Gene ID
100761174
Gene Name
proteasome (prosome, macropain) assembly chaperone 2
Gene Aliases
I79_012105
Chromosome Location
Chr.Un|NW_003614106: 730022 - 739807 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007645440.1 XP_007643630.1 1 - 2 143 108
XM_007645441.1 XP_007643631.1 1 - 2 146 108
GenBank mRNA JI885460.1 - 1 - 2 91 108

View Full Product Details