Assay Design



Gene Transcripts

Gene Symbol
Inpp1
Entrez Gene ID
100755639
Gene Name
inositol polyphosphate-1-phosphatase
Gene Aliases
I79_025102
Chromosome Location
Chr.Un|NW_003616321: 115695 - 134394 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654257.1 XP_007652447.1 1 - 2 481 85

View Full Product Details