Assay Design



Gene Transcripts

Gene Symbol
Pax1
Entrez Gene ID
100769445
Gene Name
paired box 1
Gene Aliases
I79_001851
Chromosome Location
Chr.Un|NW_003613623: 276314 - 286700 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496473.1 XP_003496521.1 2 - 3 156 91

View Full Product Details