Assay Design



Gene Transcripts

Gene Symbol
Ptplad2
Entrez Gene ID
100774830
Gene Name
protein tyrosine phosphatase-like A domain containing 2
Gene Aliases
I79_002592
Chromosome Location
Chr.Un|NW_003613640: 3228558 - 3258016 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007607660.1 XP_007605850.1 1 - 2 52 102

View Full Product Details