Assay Design



Gene Transcripts

Gene Symbol
Clic6
Entrez Gene ID
100757995
Gene Name
chloride intracellular channel 6
Gene Aliases
I79_012455
Chromosome Location
Chr.Un|NW_003614135: 127455 - 167354 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505283.2 XP_003505331.1 2 - 3 1190 82

View Full Product Details