Assay Design



Gene Transcripts

Gene Symbol
Phpt1
Entrez Gene ID
100761523
Gene Name
phosphohistidine phosphatase 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616323: 57240 - 59052 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514066.2 XP_003514114.1 1 - 2 590 59
GenBank mRNA JI880829.1 - 1 - 2 239 59
JP048437.1 - 1 - 2 239 59

View Full Product Details