Assay Design



Gene Transcripts

Gene Symbol
LOC103158792
Entrez Gene ID
103158792
Gene Name
probable gluconokinase
Gene Aliases
I79_025619
Chromosome Location
Chr.Un|NW_003620757: 8787 - 10640 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007607948.1 XP_007606138.1 - 200 64

View Full Product Details