Assay Design



Gene Transcripts

Gene Symbol
Fgf2
Entrez Gene ID
100757429
Gene Name
fibroblast growth factor 2 (basic)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614286: 549737 - 606159 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647307.1 XP_007645497.1 4 - 4 509 93

View Full Product Details