Assay Design



Gene Transcripts

Gene Symbol
Cers1
Entrez Gene ID
100765511
Gene Name
ceramide synthase 1
Gene Aliases
I79_005086
Chromosome Location
Chr.Un|NW_003613716: 641327 - 654209 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499101.2 XP_003499149.1 6 - 7 808 65

View Full Product Details