Assay Design



Gene Transcripts

Gene Symbol
Tef
Entrez Gene ID
100761103
Gene Name
thyrotrophic embryonic factor
Gene Aliases
I79_018569
Chromosome Location
Chr.Un|NW_003614859: 4679 - 16099 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007650938.1 XP_007649128.1 2 - 3 622 95

View Full Product Details