Assay Design



Gene Transcripts

Gene Symbol
LOC100774181
Entrez Gene ID
100774181
Gene Name
chromosome unknown open reading frame, human C2orf43
Gene Aliases
I79_008571
Chromosome Location
Chr.Un|NW_003613876: 103747 - 157274 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502097.2 XP_003502145.1 3 - 4 376 83
GenBank mRNA JI876754.1 - 3 - 4 377 83

View Full Product Details