Assay Design



Gene Transcripts

Gene Symbol
Sh2d1a
Entrez Gene ID
100757143
Gene Name
SH2 domain containing 1A
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614489: 303191 - 328608 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508143.1 XP_003508191.1 2 - 3 208 105
XM_007648763.1 XP_007646953.1 2 - 3 208 105

View Full Product Details