Assay Design



Gene Transcripts

Gene Symbol
Pglyrp3
Entrez Gene ID
100771434
Gene Name
peptidoglycan recognition protein 3
Gene Aliases
I79_019769
Chromosome Location
Chr.Un|NW_003615070: 237040 - 248479 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511290.1 XP_003511338.1 1 - 2 46 78

View Full Product Details