Assay Design



Gene Transcripts

Gene Symbol
Zfyve20
Entrez Gene ID
100767223
Gene Name
zinc finger, FYVE domain containing 20
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613584: 2484981 - 2511857 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495175.2 XP_003495223.1 5 - 6 756 97
GenBank mRNA JI893171.1 - 2 - 3 69 97
JP050547.1 - 2 - 3 69 97

View Full Product Details