Assay Design



Gene Transcripts

Gene Symbol
Twist1
Entrez Gene ID
103163738
Gene Name
twist family bHLH transcription factor 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613869: 506232 - 507585 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642601.1 XP_007640791.1 1 - 2 380 109

View Full Product Details