Assay Design



Gene Transcripts

Gene Symbol
Kctd1
Entrez Gene ID
100770054
Gene Name
potassium channel tetramerization domain containing 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613883: 577768 - 679824 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502223.2 XP_003502271.1 4 - 5 678 84

View Full Product Details