Assay Design



Gene Transcripts

Gene Symbol
Il9
Entrez Gene ID
100755564
Gene Name
interleukin 9
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613997: 234710 - 238276 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503741.1 XP_003503789.1 4 - 5 325 78

View Full Product Details