Assay Design



Gene Transcripts

Gene Symbol
St8sia3
Entrez Gene ID
100764454
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 3
Gene Aliases
I79_005544
Chromosome Location
Chr.Un|NW_003613738: 579509 - 584945 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007640215.1 XP_007638405.1 1 - 2 273 73

View Full Product Details