Assay Design



Gene Transcripts

Gene Symbol
Pmvk
Entrez Gene ID
100759992
Gene Name
phosphomevalonate kinase
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613854: 643401 - 654787 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501600.2 XP_003501648.1 3 - 4 539 92
GenBank mRNA JI894467.1 - 3 - 4 380 92
JP040453.1 - 3 - 4 380 92

View Full Product Details