Assay Design



Gene Transcripts

Gene Symbol
Akip1
Entrez Gene ID
100751192
Gene Name
A kinase (PRKA) interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614795: 418084 - 426644 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509904.2 XP_003509952.1 2 - 3 312 105
XM_007650557.1 XP_007648747.1 2 - 3 241 105
GenBank mRNA JI892296.1 - 2 - 3 296 105
JP048937.1 - 2 - 3 296 105

View Full Product Details