Assay Design



Gene Transcripts

Gene Symbol
Akip1
Entrez Gene ID
100751192
Gene Name
A kinase (PRKA) interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614795: 418084 - 426644 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509904.2 XP_003509952.1 4 - 5 507 89
XM_007650557.1 XP_007648747.1 4 - 5 436 89
XM_007650558.1 XP_007648748.1 3 - 4 356 89
GenBank mRNA JI892296.1 - 4 - 5 491 89
JP048937.1 - 4 - 5 491 89

View Full Product Details