Assay Design



Gene Transcripts

Gene Symbol
Chsy1
Entrez Gene ID
100770803
Gene Name
chondroitin sulfate synthase 1
Gene Aliases
I79_005479
Chromosome Location
Chr.Un|NW_003613734: 1506758 - 1560655 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007640163.1 XP_007638353.1 3 - 4 585 105
GenBank mRNA JI875248.1 - 1 - 2 487 106
JP046362.1 - 1 - 2 487 105

View Full Product Details