Assay Design



Gene Transcripts

Gene Symbol
Pnp
Entrez Gene ID
100757729
Gene Name
purine nucleoside phosphorylase
Gene Aliases
I79_017328
Chromosome Location
Chr.Un|NW_003614668: 326893 - 333119 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509215.2 XP_003509263.1 4 - 5 639 65
GenBank mRNA JP056140.1 - 4 - 5 548 65

View Full Product Details