Assay Design



Gene Transcripts

Gene Symbol
Epyc
Entrez Gene ID
100754407
Gene Name
epiphycan
Gene Aliases
I79_010808
Chromosome Location
Chr.Un|NW_003614008: 267430 - 298349 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644434.1 XP_007642624.1 4 - 5 711 92

View Full Product Details