Assay Design



Gene Transcripts

Gene Symbol
Lcn9
Entrez Gene ID
100759474
Gene Name
lipocalin 9
Gene Aliases
I79_021623
Chromosome Location
Chr.Un|NW_003615590: 2488 - 4360 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653205.1 XP_007651395.1 2 - 3 237 62

View Full Product Details