Assay Design



Gene Transcripts

Gene Symbol
Dusp8
Entrez Gene ID
100768862
Gene Name
dual specificity phosphatase 8
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614862: 21654 - 31666 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007650957.1 XP_007649147.1 3 - 4 627 76

View Full Product Details