Assay Design



Gene Transcripts

Gene Symbol
Prss48
Entrez Gene ID
100764364
Gene Name
protease, serine, 48
Gene Aliases
I79_011185
Chromosome Location
Chr.Un|NW_003614039: 254337 - 263603 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504178.1 XP_003504226.1 3 - 4 261 100

View Full Product Details