Assay Design



Gene Transcripts

Gene Symbol
Tnfrsf12a
Entrez Gene ID
100768739
Gene Name
tumor necrosis factor receptor superfamily, member 12A
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614574: 622225 - 623886 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508714.2 XP_003508762.1 2 - 3 245 71
GenBank mRNA JI889141.1 - 2 - 3 242 71
JP040392.1 - 2 - 3 242 71

View Full Product Details