Assay Design



Gene Transcripts

Gene Symbol
B3galtl
Entrez Gene ID
100759864
Gene Name
beta 1,3-galactosyltransferase-like
Gene Aliases
I79_002074
Chromosome Location
Chr.Un|NW_003613630: 220709 - 287267 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652246.1 XP_007650436.1 9 - 10 797 96
GenBank mRNA JP038488.1 - 9 - 10 897 96

View Full Product Details