Assay Design



Gene Transcripts

Gene Symbol
Akap4
Entrez Gene ID
100754812
Gene Name
A kinase (PRKA) anchor protein 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614190: 259401 - 270605 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505798.2 XP_003505846.1 2 - 3 124 93

View Full Product Details