Assay Design



Gene Transcripts

Gene Symbol
Ndufa2
Entrez Gene ID
100754444
Gene Name
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 2, 8kDa
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614866: 359994 - 361963 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510354.2 XP_003510402.1 1 - 2 186 85
GenBank mRNA JI875282.1 - 1 - 2 129 85
JP049825.1 - 1 - 2 129 85

View Full Product Details