Assay Design



Gene Transcripts

Gene Symbol
Txndc17
Entrez Gene ID
100755189
Gene Name
thioredoxin domain containing 17
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614234: 740278 - 742904 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506367.2 XP_003506415.2 1 - 2 233 83
GenBank mRNA JI871236.1 - 1 - 2 436 83
JP060946.1 - 1 - 2 436 83

View Full Product Details