Assay Design



Gene Transcripts

Gene Symbol
Gtsf1l
Entrez Gene ID
100772957
Gene Name
gametocyte specific factor 1-like
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614609: 542453 - 542987 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007649548.1 XP_007647738.1 1 - 2 278 78

View Full Product Details