Assay Design



Gene Transcripts

Gene Symbol
Drgx
Entrez Gene ID
100770585
Gene Name
dorsal root ganglia homeobox
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613589: 2118203 - 2148043 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495311.2 XP_003495359.1 1 - 2 30 57

View Full Product Details