Assay Design



Gene Transcripts

Gene Symbol
Abt1
Entrez Gene ID
100771876
Gene Name
activator of basal transcription 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613881: 79372 - 82952 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502155.2 XP_003502203.2 1 - 2 319 64

View Full Product Details